Learn More
Hamilton™ Dual syringe Diluter Valve
For use with ML503A and ML530B instruments.
Supplier:  Hamilton™ 35844
Catalog No. HAM35844
Only null left
Explore available promotions
Description
Description
  • Aluminum housing with PTFE-rotary plug
  • Rotation of valve connects two syringes to same fluid path
  • Enables larger dilutions
Specifications
Specifications
Rotation of Valve Connects Two Syringes to Same Fluid Path
Aluminum, PTFE
0 to 100 psi
Dual Syringe Diluter Valve
ML503 A and ML530 B Instruments
Product Suggestions
Product Suggestions