Learn More
Hamilton™ Chaney Adapter Assemb 700 800 7000/1700 1800 Series Microliter/Gastight™ Syring
Designed to work with 700, 800, 7000 series Microliter syringes and 1700 and 1800 series Gastight syringes
$107.81 - $280.60
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | |||||
|
14-819-10
|
Hamilton™
HAM14700 |
Hamilton Syringe Models 75 and 701 Series |
N/A
|
|
|||||
|
14-819-15
|
Hamilton™
HAM14725 |
Hamilton Syringe Models 702-750, 7000 and 1700 Series |
N/A
|
|
|||||
|
14-819-242
|
Hamilton™
HAM32146 |
Hamilton Syringe Models 800 and 1800 Series |
N/A
|
|
|||||
Description
Adapters are designed to give consistent and reproducible injections and also prevent the plungers from bending.
Specifications
| Reproducibility Adapter | |
| Ea. |