Learn More
Hamilton™ Chaney Adapter Assemb 700 800 7000/1700 1800 Series Microliter/Gastight™ Syring
Designed to work with 700, 800, 7000 series Microliter syringes and 1700 and 1800 series Gastight syringes
Supplier:  Hamilton™ HAM14725
 View more versions of this product
Catalog No. 14-819-15
Only null left
Explore available promotions
Description
Description
Adapters are designed to give consistent and reproducible injections and also prevent the plungers from bending.
Specifications
Specifications
Reproducibility Adapter
Ea.
Hamilton Syringe Models 702-750, 7000 and 1700 Series
Product Suggestions
Product Suggestions