missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ 700 Series Syringe, 10 μL, Sold by MilliporeSigma™ Supelco™

Includes a 2 in. Stainless Steel, Removable Needle

$559.13 - $559.13

Specifications

Material Stainless Steel (Tip)
Description 700 Series
Length (English) Needle 2 in.
Product Type Syringe
Needle Point Number 2
View More Specs

Products 1
Catalog Number Mfr. No. Connection Type Price Quantity  
Catalog Number Mfr. No. Connection Type Price Quantity  
111009435
View Documents
Merck Emd Millipore
20793
Removable
Each for $559.13
Only null left
 
Description

Description

Specifications

Specifications

Stainless Steel (Tip)
2 in.
2
Hamilton
Bevel
700 Series
Syringe
10 μL
51 mm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Hamilton is a registered trademark of Hamilton Co.