Learn More
Hamilton™ 1700 and 1000 Series Gastight™ SampleLock™ Syringes, 22 Gauge
Hamilton SampleLock syringes allow you to collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
Supplier: Hamilton™ HAM86326
Description
Hamilton SampleLock syringes allow you to collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
Syringes are available in sizes ranging from 1mL to 100mL. An easy-to-use twist valve and a positive rear stop on SampleLock prevents loss of sample and plunger blowout. Optional male luer and female luer lock adapter threads on the SampleLock valve making the syringes compatible with a multitude of connectors and fittings. Typical applications are:
- Headspace application
- Environmental sample collection and storage
- Pre-pressurization of gaseous sample for GC analysis
- Injection of internal standards through the valve port into samples
- Sample spiking
Sample Lock 1700 and 1000 Series Syringe Terminations
- Sample Lock (SL) - The sample lock incorporates an On/Off syringe valve with RN needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseous samples for GC analysis, and sample spiking. Not autoclavable.
- Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 26-22
Specifications
| Not Autoclavable | |
| 22 G | |
| 0.01 in. | |
| 0.02 in. | |
| 23 mm | |
| 0.72 mm | |
| 2 in. | |
| 100 psi | |
| 80°C | |
| 2 | |
| Ea. | |
| 51 mm | |
| Sample Lock, Removable |
| Borosilicate Glass (Barrel), PTFE (Tip), Stainless Steel (Plunger) | |
| 0.9 in. | |
| 0.41 mm | |
| 1.07 in. | |
| 27.1 mm | |
| To collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination | |
| General Purpose Syringe | |
| 176°F | |
| 50°F | |
| 10°C | |
| 25 mL | |
| Sharp, Bevel, Curved, Non-coring |