missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hafnium ICP Standard, 1000 ppm Hf in 3% HCl, Ricca Chemical
Supplier: Ricca Chemical Company PHF1KH500
Specifications
| Hafnium ICP Standard | |
| 14456-34-9 , 7647-01-0 , 14456-34-9 , 7732-18-5 | |
| 1,000ppm Hf | |
| 1 mL = 1 mg Hf | |
| CWGPOJHZSBOIPU-UHFFFAOYSA-L | |
| hafnium;dichloride;nonahydrate | |
| 131675691 |
| Natural Poly Bottle | |
| 94.0,3.34,0.22 | |
| 98.0,3.48,0.23 | |
| Cl2H18HfO9-2 | |
| O.O.O.O.O.O.O.O.O.[Cl-].[Cl-].[Hf] | |
| 411.525 |
Chemical Identifiers
| 14456-34-9 | |
| 411.525 | |
| 131675691 | |
| O.O.O.O.O.O.O.O.O.[Cl-].[Cl-].[Hf] |
| Cl2H18HfO9-2 | |
| CWGPOJHZSBOIPU-UHFFFAOYSA-L | |
| hafnium;dichloride;nonahydrate |