missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ H2DCFDA, >97%
Description
Fluorescent ROS indicator; becomes fluorescent when oxidized. Excitation wavelength ∽490 nm, emission ∽520 nm. Cell permeable.
Specifications
Specifications
| Quantity | 100 mg |
| Solubility | Soluble to 100mM in DMSO and to 10mM in ethanol |
| Product Line | Tocris |
| Formula Weight | 487.29 |
| Purity | 95% |
| Molecular Weight (g/mol) | 487.285 |
| Chemical Name or Material | 2-[3,6-Bis(acetyloxy)-2,7-dichloro-9H-xanthen-9-yl]benzoic acid |
Safety and Handling
| Recommended Storage | Store at -20°C |