missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GW3965 hydrochloride, 95%

Catalog No. AC468011000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468011000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468011000 Supplier Thermo Scientific™ Supplier No. 468011000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material GW3965 hydrochloride
CAS 405911-17-3
Synonym 3-[3-[N-(2-Chloro-3-trifluoromethylbenzyl)-(2,2-diphenylethyl)amino]propyloxy]phenylacetic acid hydrochloride
Quantity 100 mg
Molecular Formula C33 H31 Cl F3 N O3 . HCl
Formula Weight 618.52
Physical Form Solid
Product Type Reagent
Purity 95%