missing translation for 'onlineSavingsMsg'
Learn More

Guardian™ CP-13-11, Chemical Resistant Smooth Finish 13 mil Butyl Gloves

Butyl gloves offer protection against variety of aggressive/hazardous threats from chemicals, biologics, and radioactive materials. Ergonomic curved hand style is comfortable, with a smooth finish for enhanced grip capabilities.

$72.55 - $109.18

Specifications

Color Black
Latex Free Yes
Material Butyl
Thickness (English) 13 mil
Thickness (Metric) 0.33 mm
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Qty  
Catalog Number Mfr. No. Size Price Qty  
19-321-189
View Documents
Guardian™
51501
X-Small
Pair for $72.55
Only null left
 
19-321-190
View Documents
Guardian™
51502
Small
Pair for $89.43
Only null left
 
19321191
View Documents
Guardian™
GGL51503
Medium
Pair for $89.43
Only null left
 
19-321-192
View Documents
Guardian™
51504
Large
Pair for $109.18
Only null left
 
19-321-193
View Documents
Guardian™
51505
X-Large
Pair for $89.43
Only null left
 
Description

Description

  • For use with aldehydes, ketones, esters, and concentrated mineral acids
  • Resistant to oxidation, ozone corrosion, and abrasion
  • Shorter, thinner format than other Guardian™ Butyl Gloves
  • Rinse clean and hang dry for reuse
  • Made in the USA
Specifications

Specifications

Black
Butyl
0.33 mm
Smooth
11 in.
Yes
13 mil
Beaded
Unlined
27.9 cm
Videos
SDS
Documents

Documents