missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guardian™ CP-13-11, Chemical Resistant Smooth Finish 13 mil Butyl Gloves
Butyl gloves offer protection against variety of aggressive/hazardous threats from chemicals, biologics, and radioactive materials. Ergonomic curved hand style is comfortable, with a smooth finish for enhanced grip capabilities.
Supplier: Guardian™ 51505
Description
- For use with aldehydes, ketones, esters, and concentrated mineral acids
- Resistant to oxidation, ozone corrosion, and abrasion
- Shorter, thinner format than other Guardian™ Butyl Gloves
- Rinse clean and hang dry for reuse
- Made in the USA
Specifications
| Black | |
| Butyl | |
| 13 mil | |
| Beaded | |
| Unlined | |
| 11 in. |
| Yes | |
| X-Large | |
| 0.33 mm | |
| Smooth | |
| 51505 | |
| 27.9 cm |