missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Guanidine hydrochloride, Molecular Biology Grade

A protein denaturant | CAS: 50-01-1 | CH6ClN3 | 95.53 g/mol

Supplier:  Thermo Scientific Chemicals J6566136

 View more versions of this product

Catalog No. AAJ6566136

Only null left
Explore available promotions

Description

Description

Guanidine hydrochloride is used for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary. Guanidine hydrochloride is used in RNA isolation to dissociate nucleoproteins and inhibit RNase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanidine hydrochloride is used for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary. Guanidine hydrochloride is used in RNA isolation to dissociate nucleoproteins and inhibit RNase.

Solubility
Freely soluble in water

Notes
Protect from heat. Store away from oxidizing agents.
Specifications

Specifications

Guanidine hydrochloride
178°C to 185°C
1.34
MFCD00013026
3591990
144562
Freely soluble in water
C(=N)(N)N.Cl
95.53
CHEBI:32735
Molecular Biology
50-01-1
White
CH6ClN3
500 g
Hygroscopic
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PJJJBBJSCAKJQF-UHFFFAOYSA-N
guanidine;hydrochloride
5742
95.53
Crystalline Solid

Chemical Identifiers

50-01-1
95.53
PJJJBBJSCAKJQF-UHFFFAOYSA-N
5742
guanidine;hydrochloride
CH6ClN3
MFCD00013026
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
CHEBI:32735
C(=N)(N)N.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes serious eye irritation, May form combustible dust concentrations in air, Causes skin irritation, Harmful if swallowed or if inhaled

Serious eye damage/eye irritation (category 2), Skin corrosion/irritation (category 2), Acute toxicity (category 4)

EINECSNumber : 200-002-3

RTECSNumber : MF4300000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only