missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Guanidine hydrochloride, Molecular Biology Grade

Catalog No. p-7043128
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6566122 100 g
AAJ6566136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6566122 Supplier Thermo Scientific Chemicals Supplier No. J6566122
Only null left

A protein denaturant | CAS: 50-01-1 | CH6ClN3 | 95.53 g/mol

Guanidine hydrochloride is used for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary. Guanidine hydrochloride is used in RNA isolation to dissociate nucleoproteins and inhibit RNase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanidine hydrochloride is used for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary. Guanidine hydrochloride is used in RNA isolation to dissociate nucleoproteins and inhibit RNase.

Solubility
Freely soluble in water

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 178°C to 185°C
Color White
Density 1.34
Quantity 100 g
Beilstein 3591990
Sensitivity Hygroscopic
Merck Index 144562
Solubility Information Freely soluble in water
Formula Weight 95.53
Grade Molecular Biology
Physical Form Crystalline Solid
Chemical Name or Material Guanidine hydrochloride
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 2), Skin corrosion/irritation (category 2), Acute toxicity (category 4)
Hazard Statement Causes serious eye irritation, May form combustible dust concentrations in air, Causes skin irritation, Harmful if swallowed or if inhaled
EINECSNumber 200-002-3
RTECSNumber MF4300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only