missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Guanidine Hydrochloride, ≥99%, Calbiochem™,
$528.00 - $1,940.00
Chemical Identifiers
| CAS | 50-01-1 |
|---|---|
| Molecular Formula | CH6ClN3 |
| Molecular Weight (g/mol) | 95.53 |
| InChI Key | PJJJBBJSCAKJQF-UHFFFAOYSA-N |
| Synonym | Guanidinium Chloride |
| PubChem CID | 5742 |
| ChEBI | CHEBI:32735 |
| IUPAC Name | guanidine;hydrochloride |
| SMILES | C(=N)(N)N.Cl |
Chemical Identifiers
| 50-01-1 | |
| 95.53 | |
| Guanidinium Chloride | |
| CHEBI:32735 | |
| C(=N)(N)N.Cl |
| CH6ClN3 | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| 5742 | |
| guanidine;hydrochloride |
Specifications
| White | |
| CH6ClN3 | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| guanidine;hydrochloride | |
| 5742 | |
| ≥99% (Titration) |
| 50-01-1 | |
| Guanidinium Chloride | |
| C(=N)(N)N.Cl | |
| 95.53 | |
| CHEBI:32735 |