missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine-HCl, Molecular Grade, Promega™
Used for the isolation of intact mRNA from tissues or cultured cells.
Supplier: Promega Corporation H5383
Description
- Quality tested and certified free of DNase, RNase and protease activity
Specifications
| Guanidine-HCl | |
| 50-01-1 | |
| CH6ClN3 | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| 95.53 | |
| CHEBI:32735 | |
| Molecular |
| For the isolation of intact mRNA from tissues or cultured cells | |
| Colorless or White | |
| 500 g | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| 5742 | |
| 95.53 | |
| Fine Crystals |
Chemical Identifiers
| 50-01-1 | |
| 95.53 | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| CHEBI:32735 |
| CH6ClN3 | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| 5742 |
Safety and Handling
Recommended Storage : Store at 22° to 25°C.