missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine-HCl, Molecular Grade, Promega™
Used for the isolation of intact mRNA from tissues or cultured cells.
$77.73 - $321.49
Chemical Identifiers
| CAS | 50-01-1 |
|---|---|
| Molecular Formula | CH6ClN3 |
| Molecular Weight (g/mol) | 95.53 |
| InChI Key | PJJJBBJSCAKJQF-UHFFFAOYSA-N |
| Synonym | guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride |
| PubChem CID | 5742 |
| ChEBI | CHEBI:32735 |
Description
- Quality tested and certified free of DNase, RNase and protease activity
Chemical Identifiers
| 50-01-1 | |
| 95.53 | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| CHEBI:32735 |
| CH6ClN3 | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| 5742 |
Specifications
| 50-01-1 | |
| CH6ClN3 | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| 95.53 | |
| CHEBI:32735 | |
| Molecular | |
| Guanidine-HCl, For the isolation of intact mRNA from tissues or cultured cells |
| Colorless or White | |
| 100 g | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| 5742 | |
| 95.53 | |
| Fine Crystals |
Safety and Handling
Recommended Storage : Store at 22° to 25°C.