missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanfacine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America G04141G
Specifications
| Guanfacine Hydrochloride | |
| 213°C | |
| C9H10Cl3N3O | |
| 1 g | |
| guanfacine hydrochloride, guanfacine hcl, tenex, estulic, intuniv, n-carbamimidoyl-2-2,6-dichlorophenyl acetamide hydrochloride, guanfacin hydrochloride, connexyn, n-amidino-2-2,6-dichlorophenyl acetamide hydrochloride | |
| C1=CC(=C(C(=C1)Cl)CC(=O)N=C(N)N)Cl.Cl | |
| 282.549 | |
| 282.55 | |
| Crystalline Powder |
| 29110-48-3 | |
| White | |
| MFCD00798230 | |
| 2811 | |
| DGFYECXYGUIODH-UHFFFAOYSA-N | |
| N-(diaminomethylidene)-2-(2,6-dichlorophenyl)acetamide;hydrochloride | |
| 71401 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 29110-48-3 | |
| 282.549 | |
| DGFYECXYGUIODH-UHFFFAOYSA-N | |
| 71401 | |
| C1=CC(=C(C(=C1)Cl)CC(=O)N=C(N)N)Cl.Cl |
| C9H10Cl3N3O | |
| MFCD00798230 | |
| guanfacine hydrochloride, guanfacine hcl, tenex, estulic, intuniv, n-carbamimidoyl-2-2,6-dichlorophenyl acetamide hydrochloride, guanfacin hydrochloride, connexyn, n-amidino-2-2,6-dichlorophenyl acetamide hydrochloride | |
| N-(diaminomethylidene)-2-(2,6-dichlorophenyl)acetamide;hydrochloride |
Safety and Handling
RTECSNumber : CY1481000
TSCA : No