missing translation for 'onlineSavingsMsg'
Learn More

Guanfacine Hydrochloride 98.0+%, TCI America™

Catalog No. G0414100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0414100MG 100 mg
G04141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0414100MG Supplier TCI America Supplier No. G0414100MG

Chemical Identifiers

CAS 29110-48-3
Molecular Formula C9H10Cl3N3O
Molecular Weight (g/mol) 282.549
MDL Number MFCD00798230
InChI Key DGFYECXYGUIODH-UHFFFAOYSA-N
Synonym guanfacine hydrochloride, guanfacine hcl, tenex, estulic, intuniv, n-carbamimidoyl-2-2,6-dichlorophenyl acetamide hydrochloride, guanfacin hydrochloride, connexyn, n-amidino-2-2,6-dichlorophenyl acetamide hydrochloride
PubChem CID 71401
IUPAC Name N-(diaminomethylidene)-2-(2,6-dichlorophenyl)acetamide;hydrochloride
SMILES C1=CC(=C(C(=C1)Cl)CC(=O)N=C(N)N)Cl.Cl

Specifications

Melting Point 213°C
Color White
Quantity 100 mg
UN Number 2811
Formula Weight 282.55
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Guanfacine Hydrochloride
RTECSNumber CY1481000
TSCA No