missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ GSK 269962

Catalog No. 4009500
Encompass
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4009500 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4009500 Supplier R&D Systems™ Supplier No. 400950

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Potent and selective ROCK inhibitor

Potent Rho kinase (ROCK) inhibitor (IC50 values are 1.6 and 4 nM for recombinant human ROCK1 and ROCK2 respectively). Displays greater than 30-fold selectivity for ROCK against a panel of serine/threonine kinases. Induces vasorelaxation in preconstricted rat aorta (IC50 = 35 nM); lowers blood pressure in a rat model of hypertension.

Chemical Identifiers

CAS 850664-21-0
Molecular Formula C29H30N8O5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 570.6
Purity >99%
Molecular Weight (g/mol) 570.61
Chemical Name or Material GSK 269962
Recommended Storage Store at +4°C