missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 2193874

Catalog No. 51065
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51065 5 mg
510625 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 51065 Supplier Tocris Bioscience™ Supplier No. 5106/5
Only null left

Potent and selective TRPV4 antagonist; orally active

GSK 2193874 is a potent and selective TRPV4 antagonist (IC50 values are 2 and 40 nM for rat and human receptors, respectively); inhibits Ca2+ influx through TRPV4 channels. Prevents and reverses pulmonary edema after myocardial infarction in vivo models. Selective over a panel of ∽200 human receptors, channels and enzymes. Orally active.

Specifications

Chemical Name or Material 3-([1,4'-Bipiperidin]-1'-ylmethyl)-7-bromo-N-(1-phenylcyclopropyl)-2-[3-(trifluoromethyl)phenyl]-4-quinolinecarboxamide
CAS 1336960-13-4
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C37H38BrF3N4O
Formula Weight Observed MW: 691.62