missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 0660

Catalog No. 343350
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
343350 50 mg
343310 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 343350 Supplier Tocris Bioscience™ Supplier No. 3433/50

Selective PPARδ antagonist

Selective PPARδ antagonist (IC50 values are 0.155, > 10 and ≥ 10 μM at PPARδ, PPARα and PPARγ respectively). Exhibits inverse agonist effects when administered by itself.

Chemical Identifiers

CAS 1014691-61-2
Molecular Formula C19H18N2O5S2

Specifications

Quantity 50 mg
Formula Weight 418.49
Purity >99%
Molecular Weight (g/mol) 418.48
Chemical Name or Material GSK 0660
Recommended Storage Store at +4°C