missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSC Recombinant Rabbit Monoclonal Antibody (23GB6470)

Rabbit Recombinant Monoclonal Antibody
$656.94 - $1,779.35
Specifications
| Antigen | GSC |
|---|---|
| Clone | 23GB6470 |
| Concentration | 0.575 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
Description
GSC Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
This gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins. The encoded protein acts as a transcription factor and may be autoregulatory. A similar protein in mice plays a role in craniofacial and rib cage development during embryogenesis.Specifications
| GSC | |
| 0.575 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P56915, Q02591 | |
| 145258, 14836, 317715 | |
| A synthesized peptide derived from human GSC (1-37AA). | |
| Antibody |
| 23GB6470 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| GSC | |
| GCS; goosecoid; goosecoid homeobox; goosecoid protein; GSC; Homeobox protein goosecoid; SAMS | |
| GSC | |
| Primary | |
| Affinity chromatography |