missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GSC Recombinant Rabbit Monoclonal Antibody (23GB6470)
GREENER_CHOICE

Catalog No. MA553962
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553962 100 μL
MA553963 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553962 Supplier Invitrogen™ Supplier No. MA553962

Rabbit Recombinant Monoclonal Antibody

GSC Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins. The encoded protein acts as a transcription factor and may be autoregulatory. A similar protein in mice plays a role in craniofacial and rib cage development during embryogenesis.

Specifications

Antigen GSC
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6470
Concentration 0.575 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GSC
Gene Accession No. P56915, Q02591
Gene Alias GCS; goosecoid; goosecoid homeobox; goosecoid protein; GSC; Homeobox protein goosecoid; SAMS
Gene Symbols GSC
Host Species Rabbit
Immunogen A synthesized peptide derived from human GSC (1-37AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145258, 14836, 317715
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less