missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One VACUETTE™ CAT Serum Clot Activator Blood Collection Tubes
VACUETTE™ CAT Serum Clot Activator tubes, used for determinations in serum, are coated with micronized silica particles that activate clotting when the tubes are gently inverted.
$24.52 - $32.45
Specifications
| Color | Transparent |
|---|---|
| Product Type | Blood Collection Tube |
| Additive | Clot Activator |
| Material | PET |
| Sterility | Sterile |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
22040007
|
Greiner Bio-One
454236 |
2 mL |
Pack of 50 Each for $24.52
Case of 24 Pack for $588.48
|
|
|||||
|
22040008
|
Greiner Bio-One
454204 |
4 mL |
Pack of 50 Each for $24.52
Case of 24 Pack for $588.48
|
|
|||||
|
22040074
|
Greiner Bio-One
456089 |
6 mL |
Pack of 50 Each for $26.38
Case of 24 Pack for $633.12
|
|
|||||
|
22040009
|
Greiner Bio-One
455092 |
9 mL |
Pack of 50 Each for $32.45
Case of 24 Pack for $778.80
|
|
|||||
Description
- VACUETTE™ Blood Collection Tubes are evacuated for easy, safe sample collection. The amount of vacuum in each tube corresponds to the tube’s blood volume specification and helps ensure the correct blood-to-additive ratio.
- Injection molded from polyethylene terephthalate (PET) plastic, VACUETTE™ Blood Collection Tubes are highly transparent and virtually unbreakable.
- VACUETTE™ Serum tubes include a clot-activating additive – micronized silica particles – that is spray-dried on the interior tube wall for even disbursement and optimal blood/additive mixing.
- Optimal fill marks on every tube label indicate the proper draw volume for correct blood-to-additive ratios (within ±10% of the stated nominal volume in accordance with ISO and CLSI).
- Featuring red VACUETTE™ SAFETY Caps, designed to minimize the spraying of aerosols during decapping and color-coded to aid first-glance tube recognition.
- Sterile and latex-free.
Specifications
| Transparent | |
| Clot Activator | |
| Sterile | |
| 13 mm |
| Blood Collection Tube | |
| PET | |
| Non-ridged Pull Cap |