missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One VACUETTE™ CAT Serum Clot Activator Blood Collection Tubes

Catalog No. 22040074
Change view
Click to view available options
Capacity (Metric):
2 mL
4 mL
6 mL
9 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22040074 6 mL
22040007 2 mL
22040008 4 mL
22040009 9 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22040074 Supplier Greiner Bio-One Supplier No. 456089
Only null left

VACUETTE™ CAT Serum Clot Activator tubes, used for determinations in serum, are coated with micronized silica particles that activate clotting when the tubes are gently inverted.

  • VACUETTE™ Blood Collection Tubes are evacuated for easy, safe sample collection. The amount of vacuum in each tube corresponds to the tube’s blood volume specification and helps ensure the correct blood-to-additive ratio.
  • Injection molded from polyethylene terephthalate (PET) plastic, VACUETTE™ Blood Collection Tubes are highly transparent and virtually unbreakable.
  • VACUETTE™ Serum tubes include a clot-activating additive – micronized silica particles – that is spray-dried on the interior tube wall for even disbursement and optimal blood/additive mixing.
  • Optimal fill marks on every tube label indicate the proper draw volume for correct blood-to-additive ratios (within ±10% of the stated nominal volume in accordance with ISO and CLSI).
  • Featuring red VACUETTE™ SAFETY Caps, designed to minimize the spraying of aerosols during decapping and color-coded to aid first-glance tube recognition.
  • Sterile and latex-free.

Specifications

Color Transparent
Product Type Blood Collection Tube
Additive Clot Activator
Material PET
Sterility Sterile
Closure Type Non-ridged Pull Cap
Capacity (Metric) 6 mL
Diameter (Metric) 13 mm
Closure Color Red with Black Ring
Tube Size 13 x 100 mm
Length (Metric) 100 mm
Show More Show Less