Learn More
Greiner Bio-One VACUETTE™ CAT Serum Clot Activator Blood Collection Tubes
VACUETTE™ CAT Serum Clot Activator tubes, used for determinations in serum, are coated with micronized silica particles that activate clotting when the tubes are gently inverted.
Supplier:  Greiner Bio-One 454204
 View more versions of this product
Catalog No. 22040008
Only null left
Description
Description
  • VACUETTE™ Blood Collection Tubes are evacuated for easy, safe sample collection. The amount of vacuum in each tube corresponds to the tube’s blood volume specification and helps ensure the correct blood-to-additive ratio.
  • Injection molded from polyethylene terephthalate (PET) plastic, VACUETTE™ Blood Collection Tubes are highly transparent and virtually unbreakable.
  • VACUETTE™ Serum tubes include a clot-activating additive – micronized silica particles – that is spray-dried on the interior tube wall for even disbursement and optimal blood/additive mixing.
  • Optimal fill marks on every tube label indicate the proper draw volume for correct blood-to-additive ratios (within ±10% of the stated nominal volume in accordance with ISO and CLSI).
  • Featuring red VACUETTE™ SAFETY Caps, designed to minimize the spraying of aerosols during decapping and color-coded to aid first-glance tube recognition.
  • Sterile and latex-free.
Specifications
Chemical Identifiers
Transparent
Clot Activator
Sterile
13 mm
Blood Collection Tube
PET
Non-ridged Pull Cap
Specifications
Transparent
Clot Activator
Sterile
4 mL
Red with Black Ring
75 mm
Blood Collection Tube
PET
Non-ridged Pull Cap
13 mm
13 x 75 mm
Product Suggestions
Product Suggestions