missing translation for 'onlineSavingsMsg'
Learn More

Granisetron Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America G0401200MG

 View more versions of this product

Catalog No. G0401200MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Granisetron Hydrochloride
292°C
C18H25ClN4O
200 mg
QYZRTBKYBJRGJB-IODNYQNNSA-N
1-methyl-N-[(1S,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-yl]indazole-3-carboxamide;hydrochloride
49799997
≥98.0% (HPLC,T)
107007-99-8
White
MFCD01747034
kytril, granisetron hydrochloride, granisertron hydrochloride
CN1C2CCCC1CC(C2)NC(=O)C3=NN(C4=CC=CC=C43)C.Cl
348.875
348.88
Crystalline Powder

Chemical Identifiers

107007-99-8
348.875
QYZRTBKYBJRGJB-IODNYQNNSA-N
49799997
CN1C2CCCC1CC(C2)NC(=O)C3=NN(C4=CC=CC=C43)C.Cl
C18H25ClN4O
MFCD01747034
kytril, granisetron hydrochloride, granisertron hydrochloride
1-methyl-N-[(1S,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-yl]indazole-3-carboxamide;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : NK7882200

TSCA : No

SDS
Documents

Documents

Promotions

Promotions