missing translation for 'onlineSavingsMsg'
Learn More

Granisetron Hydrochloride 98.0+%, TCI America™

Catalog No. p-7130258
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0401200MG 200 mg
G04011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0401200MG Supplier TCI America Supplier No. G0401200MG

Chemical Identifiers

CAS 107007-99-8
Molecular Formula C18H25ClN4O
Molecular Weight (g/mol) 348.875
MDL Number MFCD01747034
InChI Key QYZRTBKYBJRGJB-IODNYQNNSA-N
Synonym kytril, granisetron hydrochloride, granisertron hydrochloride
PubChem CID 49799997
IUPAC Name 1-methyl-N-[(1S,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-yl]indazole-3-carboxamide;hydrochloride
SMILES CN1C2CCCC1CC(C2)NC(=O)C3=NN(C4=CC=CC=C43)C.Cl

Specifications

Melting Point 292°C
Color White
Quantity 200 mg
Formula Weight 348.88
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Granisetron Hydrochloride
RTECSNumber NK7882200
TSCA No