Learn More
MilliporeSigma™ Supelco™ Graduated Screw Top Vials, PTFE/Silicone Septum
Pre-assembled, clear glass vials are supplied with a PTFE/Silicone septum
Supplier:  MilliporeSigma™ Supelco™ 27520U
 View more versions of this product
Catalog No. 111009589
Only null left
Description
Description
Specifications
Chemical Identifiers
Black
Clear
PTFE, Silicone
100/Pk.
Open Top Cap
Screw Thread Vial
Pre-assembled
Glass
Yes
Phenolic
Autoclavable
Specifications
Black
Clear
Glass
PTFE, Silicone
100/Pk.
Open Top Cap
85 mm
Pre-assembled
23 mm
22 mL
Yes
Phenolic
Autoclavable
Screw Thread Vial
Product Suggestions
Product Suggestions