missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ GR-2 Septa

Designed for routine, isothermal use

Specifications

Color Gray
Material Silicone Rubber
For Use With (Application) Routine, Isothermal Use
Temperature (Metric) Max 200°C
View More Specs

Products 6
Catalog Number Mfr. No. Diameter (Metric) Outer Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Outer Quantity Price Quantity & Availability  
111011148
View Documents Promotion Details
MilliporeSigma™ Supelco™
SGMA20405
9.5 mm 100 Pk. N/A
Only null left
 
111010227
View Documents Promotion Details
MilliporeSigma™ Supelco™
20625
9.5 mm 250 Pk. N/A
Only null left
 
111020757
View Documents Promotion Details
MilliporeSigma™ Supelco™
20627
9.5 mm 1000 Pk. N/A
Only null left
 
111011152
View Documents Promotion Details
MilliporeSigma™ Supelco™
20441
10 mm 100 Pk. N/A
Only null left
 
111011151
View Documents Promotion Details
MilliporeSigma™ Supelco™
SGMA20421
11 mm 100 Pk. N/A
Only null left
 
111011150
View Documents Promotion Details
MilliporeSigma™ Supelco™
20413
12.5 mm 100 Pk. N/A
Only null left
 
Description

Description

  • Septa are very important components of a syringe injection system
  • Typically fabricated from elastomeric, self-sealing materials such as silicone rubber, neoprene, or fluoroelastomers
  • Suitable to conveniently introducing the sample without causing the system to leak
  • Improper septa decompose under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram
  • These low cost, gray silicone rubber septa are designed for routine, isothermal use
  • Inlet Temperature Range: 50°C to 200°C
Specifications

Specifications

Gray
Silicone Rubber
Routine, Isothermal Use
200°C
Videos
SDS
Documents

Documents