missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ GR-2 Septa

Catalog No. 111011148
Change view
Click to view available options
Diameter (Metric) Outer:
9.5 mm
10 mm
11 mm
12.5 mm
Quantity:
100 Pk.
1000 Pk.
250 Pk.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer Quantity
111011148 9.5 mm 100 Pk.
111010227 9.5 mm 250 Pk.
111020757 9.5 mm 1000 Pk.
111011152 10 mm 100 Pk.
111011151 11 mm 100 Pk.
111011150 12.5 mm 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 111011148 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMA20405

Designed for routine, isothermal use

  • Septa are very important components of a syringe injection system
  • Typically fabricated from elastomeric, self-sealing materials such as silicone rubber, neoprene, or fluoroelastomers
  • Suitable to conveniently introducing the sample without causing the system to leak
  • Improper septa decompose under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram
  • These low cost, gray silicone rubber septa are designed for routine, isothermal use
  • Inlet Temperature Range: 50°C to 200°C

Specifications

Type GR-2 Septum
Temperature (Metric) Max 200°C
Material Silicone Rubber
Diameter (English) Outer 3/8 in.
Diameter (Metric) Outer 9.5 mm
For Use With (Application) Routine, Isothermal Use
Color Gray
Quantity 100 Pk.