missing translation for 'onlineSavingsMsg'
Learn More

Promega GoTaq™ G2 Hot Start Green Master Mixes

Catalog No. p-5379091
Encompass
Change view
Click to view available options
No. of Reactions:
100 reactions
1000 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PRM7422 100 reactions
PRM7423 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRM7422 Supplier Promega Supplier No. M7422
Only null left

GoTaq™ G2 Hot Start Master Mixes are ready-to-use mixes containing GoTaq™ G2 Hot Start Polymerase, buffer, dNTPs and optimized magnesium

  • Room-Temperature PCR Setup with High-Performance Master Mix
  • All-in-one master mix: just add template and primers
  • Eliminate nonspecific amplification with a hot start enzyme
  • Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required

Specifications

Content And Storage −30° to −10°C
No. of Reactions 100 reactions
Sample Type Green master mix