missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GoTaq™ G2 Hot Start Green Master Mixes
Description
- Room-Temperature PCR Setup with High-Performance Master Mix
- All-in-one master mix: just add template and primers
- Eliminate nonspecific amplification with a hot start enzyme
- Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required
Specifications
Specifications
| Content And Storage | −30° to −10°C |
| No. of Reactions | 100 reactions |
| Sample Type | Green master mix |