Learn More
Corning™ 100x15mm Petri Dish with three vents, Optimized for automation
GREENER_CHOICE
Catalog No. 07202031
Change view
Click to view available options
Sterility:
Aseptically Manufactured
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
07202031 Sterile
07202030 Aseptically Manufactured
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07202031
Edge
Optimized for automation.
Gosselin 100 x 15 mm Petri dishes have been designed in collaboration with Petri dish filling equipment manufacturers . These Petri dishes are ideal for microbiological examination either by surface plating or plate pouring techniques.
  • Crystal polystyrene
  • Three vents
  • Optimal flatness
  • Manufactured in class 100
  • SAL 10-6
  • Easy-opening sleeves
Specifications
Material Polystyrene
Shape Round
Sterility Sterile
For Use With (Application) Optimized for automation
Vent 3
Height (Metric) 14.2 mm
Diameter (Metric) 90 mm