Learn More
Promega GoScript Reverse Transcription Mix, Oligo(dT), 100 reactions
GoScript Reverse Transcriptase utilizes M-MLV and state-of-the-art buffer technology designed for qPCR to deliver robust, reliable cDNA synthesis of a full range of rare and abundant transcripts, even in the presence of inhibitors.
Supplier:  Promega A2791
Encompass
Catalog No. PRA2791
Only null left
Description
Description
  • Reliable cDNA Synthesis for Sensitive Detection of Expression
  • Available as a standalone enzyme, a complete reverse transcription kit or as a master mix with Oligo(dT) or Random Primers. RNasin(R) RNase Inhibitor protection included with Kit and Mixes
  • Sensitive transcription of both high−copy and low−copy messages
  • Transcribes short and long transcripts; processes through secondary structure
Specifications
Specifications
-30C TO -10C
100 reactions
Product Suggestions
Product Suggestions