missing translation for 'onlineSavingsMsg'
Learn More

Gold Standard For ICP, 1000μg/mL in 10% HCl, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLAU32M

Catalog No. PLAU32M


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Au1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Gold Standard
7440-57-5
Au
AA, ICP
Certified Reference Material
[Au]
gold
30 mL
CHEBI:30050
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00003436
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
PCHJSUWPFVWCPO-UHFFFAOYSA-N
10% HCl
196.97
23985

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
CHEBI:30050
[Au]
Au
MFCD00003436
23985
gold
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

May cause cancer. Causes skin irritation, serious eye irritation

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only