Learn More
Gold AA Standard, Certified, 1000 ppm (1mL = 1mg Au), LabChem™
Supplier: LabChem LC148901
Specifications
| Gold AA Standard | |
| Amber Glass | |
| 89.82,9.98,0.2 | |
| Liquid | |
| 1000 ppm (1mL = 1mg Au) | |
| UN3264 | |
| Soluble in water | |
| Cl.Cl[Au](Cl)Cl | |
| 339.77 | |
| 393.83 |
| 1000ppm (1mL = 1mg Au) | |
| 16961-25-4,7647-01-0,7732-18-5 | |
| Passes Test | |
| AuCl4H | |
| HAuCl4·3H2O | |
| Yellow | |
| VDLSFRRYNGEBEJ-UHFFFAOYSA-K | |
| trichlorogold hydrochloride | |
| 500 mL | |
| Certified |
Chemical Identifiers
| 16961-25-4 | |
| 339.77 | |
| trichlorogold hydrochloride |
| AuCl4H | |
| VDLSFRRYNGEBEJ-UHFFFAOYSA-K | |
| Cl.Cl[Au](Cl)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature