missing translation for 'onlineSavingsMsg'
Learn More

Gnetol 97.0+%, TCI America™

Catalog No. G0371100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0371100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G0371100MG Supplier TCI America Supplier No. G0371100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86361-55-9
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD12405548
InChI Key DQULNTWGBBNZSC-SNAWJCMRSA-N
Synonym 2,3′C,5′C,6-Tetrahydroxy-trans-stilbene
PubChem CID 45382232
IUPAC Name 2-[(E)-2-(3,5-dihydroxyphenyl)ethenyl]benzene-1,3-diol
SMILES C1=CC(=C(C(=C1)O)C=CC2=CC(=CC(=C2)O)O)O

Specifications

Melting Point 271°C
Quantity 100 mg
Formula Weight 244.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Gnetol