accessibility menu, dialog, popup

UserName

OHOHHOHO(E/Z)

CAS RN 86361-55-9

IUPAC Name: 2-[2-(3,5-dihydroxyphenyl)ethenyl]benzene-1,3-diol
Synonyms: 2-[2-(3,5-dihydroxyphenyl)ethenyl]benzene-1,3-diol
Molecular Weight (g/mol): 244.25
Molecular Formula: C14H12O4
InChi Key: DQULNTWGBBNZSC-UHFFFAOYSA-N
SMILES: OC1=CC(C=CC2=C(O)C=CC=C2O)=CC(O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results