missing translation for 'onlineSavingsMsg'
Learn More

Glyoxylic acid, 50% in water

Catalog No. AC411065000
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411065000 500 g Glass bottle
AC411060050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC411065000 Supplier Thermo Scientific Chemicals Supplier No. 411065000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 298-12-4 | C2H2O3 | 74.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 298-12-4 , 7732-18-5
Molecular Formula C2H2O3
Molecular Weight (g/mol) 74.035
MDL Number MFCD00006958
InChI Key HHLFWLYXYJOTON-UHFFFAOYSA-N
Synonym glyoxylic acid, 2-oxoacetic acid, oxoacetic acid, oxoethanoic acid, glyoxalic acid, formylformic acid, acetic acid, oxo, oxalaldehydic acid, glyoxylate, alpha-ketoacetic acid
PubChem CID 760
ChEBI CHEBI:16891
IUPAC Name oxaldehydic acid
SMILES C(=O)C(=O)O

Specifications

Color Colorless to Yellow
Boiling Point 111.0°C
Melting Point -93.0°C
Density 1.3000g/mL
Packaging Glass bottle
Refractive Index 1.4140 to 1.4180
Specific Gravity 1.3
Linear Formula HC(O)COOH
Quantity 500 g
Beilstein 03, IV, 1489
Fieser 05,320; 07,162; 09,228
Physical Form Liquid
Merck Index 15, 4546
Solubility Information Solubility in water: miscible.
Formula Weight 74.04
Percent Purity ≥50%
Chemical Name or Material Glyoxylic acid, 50% in water
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-058-5
RTECSNumber MD4550000
TSCA TSCA

RUO – Research Use Only