missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GlyH 101
Description
GlyH 101 is a reversible, voltage-dependent CFTR chloride channel blocker (Ki = 4.3 μM). Inhibits forskolin -induced hyperpolarization in nasal potential differences and inhibits cholera toxin-induced intestinal fluid secretion in mice.
Specifications
Specifications
| Chemical Name or Material | GlyH 101 |
| CAS | 328541-79-3 |
| Synonym | N-2-Naphthalenyl-2-[(3,5-dibromo-2,4-dihydroxyphenyl)methylene]glycine hydrazide |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C19H15Br2N3O3 |
| Formula Weight | 493.15 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use