missing translation for 'onlineSavingsMsg'
Learn More

Glycylglycine, 0.2M buffer soln., pH 8.5, Thermo Scientific™

$110.49 - $170.36

Chemical Identifiers

CAS 556-50-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00008130
InChI Key YMAWOPBAYDPSLA-UHFFFAOYSA-N
PubChem CID 11163
ChEBI CHEBI:17201
IUPAC Name 2-[(2-aminoacetyl)amino]acetic acid
SMILES C(C(=O)NCC(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60717AE
View Documents
Thermo Scientific Chemicals
J60717AE
100 mL
Each for $110.49
Only null left
 
AAJ60717AK
View Documents
Thermo Scientific Chemicals
J60717AK
250 mL
Each for $170.36
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

556-50-3
132.119
YMAWOPBAYDPSLA-UHFFFAOYSA-N
CHEBI:17201
C(C(=O)NCC(=O)O)N
C4H8N2O3
MFCD00008130
11163
2-[(2-aminoacetyl)amino]acetic acid

Specifications

556-50-3
0.2 M
Liquid
C4H8N2O3
MFCD00008130
C(C(=O)NCC(=O)O)N
132.119
CHEBI:17201
Colorless
8.5
100 mL
Odorless
YMAWOPBAYDPSLA-UHFFFAOYSA-N
2-[(2-aminoacetyl)amino]acetic acid
11163
Glycylglycine buffer soln.
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents