missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycyl-L-leucine 99.0+%, TCI America™
Supplier: TCI America G01815G
Specifications
| Glycyl-L-leucine | |
| 235°C | |
| C8H16N2O3 | |
| 5 g | |
| DKEXFJVMVGETOO-LURJTMIESA-N | |
| (2S)-2-[(2-aminoacetyl)amino]-4-methylpentanoic acid | |
| 92843 | |
| 188.23 | |
| Crystalline Powder |
| 869-19-2 | |
| White | |
| MFCD00008127 | |
| glycyl-l-leucine, n-glycyl-l-leucine, gly-leu, glycylleucine, s-2-2-aminoacetamido-4-methylpentanoic acid, l-leucine, glycyl, glycyl-leucine, unii-8l97v505gu, h-gly-leu-oh, chembl56119 | |
| CC(C)CC(C(=O)O)NC(=O)CN | |
| 188.227 | |
| CHEBI:73514 | |
| ≥99.0% (T) |
Chemical Identifiers
| 869-19-2 | |
| 188.227 | |
| DKEXFJVMVGETOO-LURJTMIESA-N | |
| 92843 | |
| (2S)-2-[(2-aminoacetyl)amino]-4-methylpentanoic acid |
| C8H16N2O3 | |
| MFCD00008127 | |
| glycyl-l-leucine, n-glycyl-l-leucine, gly-leu, glycylleucine, s-2-2-aminoacetamido-4-methylpentanoic acid, l-leucine, glycyl, glycyl-leucine, unii-8l97v505gu, h-gly-leu-oh, chembl56119 | |
| CHEBI:73514 | |
| CC(C)CC(C(=O)O)NC(=O)CN |
Safety and Handling
TSCA : No