missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-leucine 99.0+%, TCI America™

Catalog No. p-7130179
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01811G 1 g
G01815G 5 g
G018110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G01811G Supplier TCI America Supplier No. G01811G
Only null left

Chemical Identifiers

CAS 869-19-2
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.227
MDL Number MFCD00008127
InChI Key DKEXFJVMVGETOO-LURJTMIESA-N
Synonym glycyl-l-leucine, n-glycyl-l-leucine, gly-leu, glycylleucine, s-2-2-aminoacetamido-4-methylpentanoic acid, l-leucine, glycyl, glycyl-leucine, unii-8l97v505gu, h-gly-leu-oh, chembl56119
PubChem CID 92843
ChEBI CHEBI:73514
IUPAC Name (2S)-2-[(2-aminoacetyl)amino]-4-methylpentanoic acid
SMILES CC(C)CC(C(=O)O)NC(=O)CN

Specifications

Melting Point 235°C
Color White
Quantity 1 g
Formula Weight 188.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-L-leucine