accessibility menu, dialog, popup

UserName

H3CCH3(S)NHONH2OHO

CAS RN 869-19-2

IUPAC Name: (2S)-2-(2-aminoacetamido)-4-methylpentanoic acid
Synonyms: glycyl-L-leucine
Molecular Weight (g/mol): 188.23
Molecular Formula: C8H16N2O3
InChi Key: DKEXFJVMVGETOO-LURJTMIESA-N
SMILES: CC(C)C[C@H](NC(=O)CN)C(O)=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results