missing translation for 'onlineSavingsMsg'
Learn More

Glycine, Ultrapure, 99%, Spectrum™ Chemical

$395.21 - $9,120.04

Chemical Identifiers

CAS 56-40-6
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18610747
View Documents Promotion Details
N/A 100 g Poly Bottle
Each for $395.21
Only null left
 
18610746
View Documents Promotion Details
N/A 1 kg Poly Bottle
Each for $1,111.46
Only null left
 
18610749
View Documents Promotion Details
N/A 5 kg Poly Bottle
Each for $3,157.46
Only null left
 
18610748
View Documents Promotion Details
N/A 12 kg Poly Bottle
Each for $9,120.04
Only null left
 
Description

Description

Specifications

Chemical Identifiers

56-40-6

Specifications

0.1 (1 M Solution at 280 nm)
1
2e-05
0.002
99%
Glycine
56-40-6
7e-05
To Pass Test
6.5e-05
Ultrapure
Videos
SDS
Documents

Documents

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.