missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, Ultrapure, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor GL157100GM
| Quantity | 100 g |
|---|---|
| Packaging | Poly Bottle |
Chemical Identifiers
| 56-40-6 |
Specifications
| Glycine | |
| 56-40-6 | |
| 7e-05 | |
| To Pass Test | |
| Poly Bottle | |
| 100 g | |
| Ultrapure |
| 0.1 (1 M Solution at 280 nm) | |
| 1 | |
| 2e-05 | |
| 0.002 | |
| 6.5e-05 | |
| 99% |