missing translation for 'onlineSavingsMsg'
Learn More

Glycine n-propyl ester hydrochloride

CAS: 13049-01-9 | C5H12ClNO2 | 153.606 g/mol

$73.82 - $259.40

Chemical Identifiers

CAS 13049-01-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.606
MDL Number MFCD00151829
InChI Key YDBMNSABUROTEK-UHFFFAOYSA-N
Synonym propyl 2-aminoacetate hydrochloride, propyl aminoacetate hydrochloride, glycine n-propyl ester hydrochloride, glycine propyl ester hydrochloride, propyl glycinate hydrochloride, n-propyl glycinate hydrochloride, glycine n-propyl ester hcl, glycine, propyl ester,hydrochloride 1:1
PubChem CID 2759254
IUPAC Name propyl 2-aminoacetate;hydrochloride
SMILES CCCOC(=O)CN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH5025003
View Documents Promotion Details
N/A 1 g
Each for $73.82
Only null left
 
AAH5025006
View Documents Promotion Details
N/A 5 g
Each for $259.40
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13049-01-9
153.606
YDBMNSABUROTEK-UHFFFAOYSA-N
2759254
CCCOC(=O)CN.Cl
C5H12ClNO2
MFCD00151829
propyl 2-aminoacetate hydrochloride, propyl aminoacetate hydrochloride, glycine n-propyl ester hydrochloride, glycine propyl ester hydrochloride, propyl glycinate hydrochloride, n-propyl glycinate hydrochloride, glycine n-propyl ester hcl, glycine, propyl ester,hydrochloride 1:1
propyl 2-aminoacetate;hydrochloride

Specifications

78°C to 80°C
C5H12ClNO2
propyl 2-aminoacetate hydrochloride, propyl aminoacetate hydrochloride, glycine n-propyl ester hydrochloride, glycine propyl ester hydrochloride, propyl glycinate hydrochloride, n-propyl glycinate hydrochloride, glycine n-propyl ester hcl, glycine, propyl ester,hydrochloride 1:1
CCCOC(=O)CN.Cl
153.606
117.15
13049-01-9
MFCD00151829
YDBMNSABUROTEK-UHFFFAOYSA-N
propyl 2-aminoacetate;hydrochloride
2759254
Glycine n-propyl ester hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 235-925-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only