missing translation for 'onlineSavingsMsg'
Learn More

Glycine n-propyl ester hydrochloride

CAS: 13049-01-9 | C5H12ClNO2 | 153.606 g/mol

Supplier:  Thermo Scientific Chemicals H5025006

Quantity 5 g

 View more versions of this product

Catalog No. AAH5025006


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13049-01-9
153.606
YDBMNSABUROTEK-UHFFFAOYSA-N
2759254
CCCOC(=O)CN.Cl
C5H12ClNO2
MFCD00151829
propyl 2-aminoacetate hydrochloride, propyl aminoacetate hydrochloride, glycine n-propyl ester hydrochloride, glycine propyl ester hydrochloride, propyl glycinate hydrochloride, n-propyl glycinate hydrochloride, glycine n-propyl ester hcl, glycine, propyl ester,hydrochloride 1:1
propyl 2-aminoacetate;hydrochloride

Specifications

78°C to 80°C
13049-01-9
MFCD00151829
YDBMNSABUROTEK-UHFFFAOYSA-N
propyl 2-aminoacetate;hydrochloride
153.606
117.15
Glycine n-propyl ester hydrochloride
C5H12ClNO2
propyl 2-aminoacetate hydrochloride, propyl aminoacetate hydrochloride, glycine n-propyl ester hydrochloride, glycine propyl ester hydrochloride, propyl glycinate hydrochloride, n-propyl glycinate hydrochloride, glycine n-propyl ester hcl, glycine, propyl ester,hydrochloride 1:1
CCCOC(=O)CN.Cl
5 g
2759254
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 235-925-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions