Learn More
Glycine n-propyl ester hydrochloride
CAS: 13049-01-9 | C5H12ClNO2 | 153.606 g/mol
Supplier: Thermo Scientific Chemicals H5025003
| Quantity | 1 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13049-01-9 | |
| 153.606 | |
| YDBMNSABUROTEK-UHFFFAOYSA-N | |
| 2759254 | |
| CCCOC(=O)CN.Cl |
| C5H12ClNO2 | |
| MFCD00151829 | |
| propyl 2-aminoacetate hydrochloride, propyl aminoacetate hydrochloride, glycine n-propyl ester hydrochloride, glycine propyl ester hydrochloride, propyl glycinate hydrochloride, n-propyl glycinate hydrochloride, glycine n-propyl ester hcl, glycine, propyl ester,hydrochloride 1:1 | |
| propyl 2-aminoacetate;hydrochloride |
Specifications
| 78°C to 80°C | |
| 13049-01-9 | |
| MFCD00151829 | |
| YDBMNSABUROTEK-UHFFFAOYSA-N | |
| propyl 2-aminoacetate;hydrochloride | |
| 153.606 | |
| 117.15 |
| Glycine n-propyl ester hydrochloride | |
| C5H12ClNO2 | |
| propyl 2-aminoacetate hydrochloride, propyl aminoacetate hydrochloride, glycine n-propyl ester hydrochloride, glycine propyl ester hydrochloride, propyl glycinate hydrochloride, n-propyl glycinate hydrochloride, glycine n-propyl ester hcl, glycine, propyl ester,hydrochloride 1:1 | |
| CCCOC(=O)CN.Cl | |
| 1 g | |
| 2759254 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 235-925-0
TSCA : No
Recommended Storage : Ambient temperatures