missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine methyl ester hydrochloride, 99%
CAS: 5680-79-5 | C3H8ClNO2 | 125.55 g/mol
$80.25 - $893.11
Chemical Identifiers
| CAS | 5680-79-5 |
|---|---|
| Molecular Formula | C3H8ClNO2 |
| Molecular Weight (g/mol) | 125.55 |
| MDL Number | MFCD00012870 |
| InChI Key | COQRGFWWJBEXRC-UHFFFAOYSA-N |
| Synonym | glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride |
| PubChem CID | 122755 |
| IUPAC Name | methyl 2-aminoacetate;hydrochloride |
| SMILES | [H+].[Cl-].COC(=O)CN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA1408322
|
N/A | 100 g |
Each for $80.25
|
|
|||||
|
AAA1408336
|
N/A | 500 g |
Each for $222.84
|
|
|||||
|
AAA140830E
|
N/A | 2500 g |
Each for $893.11
|
|
|||||
Description
Glycine methyl ester hydrochloride a non-essential amino acid used for human development.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlycine methyl ester hydrochloride a non-essential amino acid used for human development.
Solubility
Soluble in water (2% w/v) and methanol (50 mg/ml).
Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
Chemical Identifiers
| C3H8ClNO2 | |
| MFCD00012870 | |
| glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride | |
| methyl 2-aminoacetate;hydrochloride |
Specifications
| ∼175°C (decomposition) | |
| C3H8ClNO2 | |
| 3593644 | |
| Soluble in water (2% w/v) and methanol (50mg/ml). | |
| [H+].[Cl-].COC(=O)CN | |
| methyl 2-aminoacetate;hydrochloride | |
| 122755 | |
| 99% |
| 5680-79-5 | |
| MFCD00012870 | |
| glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride | |
| COQRGFWWJBEXRC-UHFFFAOYSA-N | |
| 1 | |
| 125.55 | |
| 125.56 | |
| Glycine methyl ester hydrochloride |
Safety and Handling
EINECSNumber : 227-139-1
TSCA : No
RUO – Research Use Only